Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific

Supplier: Thermo Scientific
Pierce
37580 37515 37580 37517 37517 37515
PI37515EA 271.09 USD InStock
PI37515 CAPI37580 CAPI37517 PI37517 PI37580 CAPI37515
SuperBlock™ (PBS) Blocking Buffer, Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
SuperBlock™ Blocking Buffer is an optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4°C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilising them for drying and storage for later use.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty