Would you like to log in again?
Pierce™ Clear Milk Blocking Buffer (10X), Thermo Scientific
Supplier: Thermo Scientific
Pierce
37587 37587
PI37587EA 233.37 USD InStock
CAPI37587 PI37587
Pierce™ Clear Milk Blocking Buffer (10X), Thermo Scientific
Buffers Electrophoresis Buffers Blocking Buffers
Thermo Scientific Pierce Clear Milk Blocking Buffer (10X) is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.
  • Kit stable for 1 year stored at 4 °C
  • Convenient and saves storage space
  • Easy to use with ready-to-dilute solution
Clear Milk Blocking Buffer (10X) is a proprietary pre-formulated milk solution for blocking excess nonspecific binding sites, reducing background in Western blotting applications and diluting antibodies when used with nitrocellulose and PVDF membranes. Pierce Clear Milk Blocking Buffer (10X) provides lower background, enhanced sensitivity, extended shelf life and reproducible results compared to homemade buffers based on dissolved non-fat dry milk.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty