Would you like to log in again?
Pierce 1-Step™ ABTS Substrate Solution, Thermo Scientific
Supplier: Thermo Scientific
1-Step™
37615 37615
PI37615EA 177.49 USD InStock
PI37615 CAPI37615
Pierce 1-Step™ ABTS Substrate Solution, Thermo Scientific
Assays ELISAs
ABTS substrate is a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.
  • Green product has two major absorbance peaks, 410 nm and 65 0nm
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty