Would you like to log in again?
Pierce™ Protein A IgG Purification Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
44667 44667
PI44667EA 675.05 USD InStock
PI44667 CAPI44667
Pierce™ Protein A IgG Purification Kit, Thermo Scientific
Protein Purification Protein Purification Resins
The Protein A IgG Purification Kit is useful for routine purification of antibodies.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum
  • Agarose resin support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Superior manufacturing method immobilizes Protein A by charge-free, leach-resistant covalent bonds, resulting in low non specific binding Enables multiple uses without decline in yield
  • Pre-packed Protein A columns and desalting columns supplied in the kit make IgG purification easy and efficient
Samples containing IgG are incubated with immobilized Protein A in a buffer that facilitates binding. After non IgG components are washed from the column, the bound IgG is recovered by elution. The supplied buffers provide for maximum immunoglobulin binding and elution efficiency with the Protein A column.
Delivery information: Kit contains Protein A columns (5 x 1 mL), Protein A IgG Binding Buffer (1 L), IgG Elution Buffer (500 mL), and Desalting Columns (5 x 5 mL).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty