Would you like to log in again?
Pierce™ F(ab')2 Micro Preparation Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
44688 44688
PI44688EA 738.28 USD InStock
CAPI44688 PI44688
Pierce™ F(ab')2 Micro Preparation Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce F(ab')2 Micro Preparation Kit uses immobilized pepsin protease to selectively digest whole human and other IgG antibodies to make F(ab')2 fragments that retain antigen binding activity.
  • Enzyme free digestion products
  • Enhanced yield and sample purity
  • Spin format reduces sample processing time
  • Ideal when sample size is limited—Use the micro kit for 25 to 250 µg IgG.
  • Flexible, protocols are included for multiple species and IgG subclasses, as well as sample size and concentration
Suitable for human and other species of IgG. The kit procedure is optimized for human, mouse and rabbit IgG. Papain-based digestion is effective for certain other species and subclasses of IgG (e.g., goat and pig), although the purification step requires that the antibody effectively binds to Protein A. Immobilized pepsin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme.
Delivery information: Kit includes pepsin agarose (0.5 mL), F(ab')2 digestion buffer (55 mL), NAb protein A Plus spin columns, 0.2 mL (2 columns), PBS packs (each makes 500 mL) (2 packs), IgG elution buffer (50 mL), Zeba spin desalting columns, 7K MWCO, 0.5 mL (10 columns), spin columns, 0.8 mL (10 columns) and microcentrifuge tubes, 2 mL (30 tubes).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty