Would you like to log in again?
Pierce™ AminoLink™ Immobilization Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
44890 44890
PI44890EA 518.22 USD InStock
CAPI44890 PI44890
Pierce™ AminoLink™ Immobilization Kit, Thermo Scientific
Protein Purification Protein Purification Kits
The AminoLink Immobilization Kit contains AminoLink Coupling Resin and associated reagents sufficient to perform 5 protein immobilizations, with expected an expected yield of 2 to 20 mg protein per column.
  • AminoLink Coupling Resin—aldehyde-activated crosslinked, 4% beaded agarose
  • Ideal for antibodies and other proteins—immobilize molecules via primary amines (-NH2)
  • Efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer, with or without organic solvent)
  • Coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
  • Better than immobilization to CNBr-activated agarose as the bond is more stable and uncharged, resulting in less non specific binding in affinity purification procedures
  • Prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions
AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.
Proteins and other molecules with primary amines can be covalently attached (immobilized) to AminoLink Resin to make chromatography columns for use in affinity purification. The aldehyde groups form stable secondary amine bonds with primary amines such as exist in the side chain of lysine (K) residues, which are generally abundant and readily accessible in proteins. Once a protein is immobilized, the prepared affinity resin can be used for a variety of batch and column affinity purification methods involving binding interactions with the immobilized protein. The resin and linkage are stable in most binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for multiple rounds of affinity purification procedures.
The AminoLink Resin immobilization reaction involves spontaneous formation of Schiff base bonds between aldehydes and amines and their subsequent stabilization by incubation with a mild reductant.
Accessories information: The AminoLink™ reductant (sodium cyanoborohydride) promotes the formation of stable bonds between aldehyde- and amine-containing molecules, enabling efficient labeling, conjugation, and immobilization of proteins and other molecules.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty