Would you like to log in again?
Pierce™ Protein IgG Plus Orientation Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
44893 44893 44990 44990
PI44893EA 814.86 USD InStock
PI44893 CAPI44893 CAPI44990 PI44990
Pierce™ Protein IgG Plus Orientation Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ Protein IgG Plus Orientation Kits uses a crosslinker to covalently attach antibody that is bound to Protein A (or Protein G) agarose resin, thereby creating a reusable affinity column for purification of antigens.
  • Contaminant free antigen purification and less antibody leaching
  • Covalent, leak resistant antibody immobilization
  • Gentle binding and crosslinking conditions
  • Less antibody leaching
  • Crosslinked 6% beaded agarose with recombinant Protein A (or Protein G) thus making it high quality resin
  • Oriented immobilization that is able to capture passing antigen in the mobile phase without steric hindrance
Delivery information: Kit includes Recombinant Protein A [or Protein G] columns, 2 mL (2 columns), Disuccinimidyl suberate [DSS] (2 x 13 mg), PBS pack (makes 500 mL), Blocking Buffer (6 mL), IgG Elution Buffer (15 mL), Antibody Binding/Wash Buffer (120 mL), and column accessories.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty