Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ AminoLink™ Plus Immobilization Kit, 2 ml, Thermo Scientific

Supplier: Thermo Fisher Scientific
20394 20394 44894 44894
PI44894EA 571.2 USD InStock
CAPI44894 PI44894 CAPI20394 PI20394
Pierce™ AminoLink™ Plus Immobilization Kit, 2 ml, Thermo Scientific
Protein Purification Protein Purification Kits
The AminoLink Plus Immobilization Kit contains AminoLink Plus Coupling Resin and additional reagents needed to perform 5 protein immobilizations, with expected yields of 2 to 40 mg protein per column.

  • Aldehyde-activated crosslinked 4% beaded agarose
  • Immobilize molecules via primary amines (-NH2)
  • Efficient coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent)
  • Coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
  • Versatile and reusable

The coupling resin uses aldehyde-activated agarose beads for high-yield covalent coupling of antibodies (proteins) via primary amines to prepare columns for affinity purification.

Delivery information: Kit contains AminoLink™ Plus columns, 2 mL (5 columns), PBS pack (makes 500 mL), citrate-carbonate buffer pack (makes 500 mL), sodium cyanoborohydride solution, 5 M (0.5 mL), quenching buffer (60 mL), wash solution (240 mL), and column accessories.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty