Would you like to log in again?
Pierce™ CarboxyLink™ Immobilization Kit, 2 ml, Thermo Scientific
Supplier: Thermo Fisher Scientific
44899 44899
PI44899EA 837.42 USD InStock
PI44899 CAPI44899
Pierce™ CarboxyLink™ Immobilization Kit, 2 ml, Thermo Scientific
Protein Purification Protein Purification Kits
The Thermo Scientific CarboxyLink Immobilization Kit is for covalent immobilization of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.
  • Ideal for unmodified peptides, immobilizes peptides with high capacity and various orientations without steric hindrance
  • DADPA-activated crosslinked 4% beaded agarose acting as a CarboxyLink™ coupling resin
  • With stable linkage, flexible and gentle coupling conditions
  • Sufficient for Immobilizing 1 to 2 mg of peptide per column
CarboxyLink Resin is crosslinked beaded agarose that has been activated with diamino-dipropylamine (DADPA) to contain long spacer arms, each with a primary amine at the end
Because EDC crosslinks carboxylates to primary amines, peptides and other molecules that contain both of these functional groups will be variously polymerized as well as immobilized to the CarboxyLink Resin. For peptides whose only carboxyl and amino groups are at the termini, coupling with the CarboxyLink Kit will result in end-to-end chains of peptide that are linked by their C-terminal ends to the support. Generally, this structure provides for high capacity and low steric hindrance for use in affinity purification. However, in the case of peptides containing multiple primary amines (lysine) or carboxylates (glutamic and aspartic acid) in the middle of their sequence, the CarboxyLink Coupling Method may cause key binding sites to be blocked, resulting in high coupling capacity but poor binding capacity for affinity purification.
When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins can also be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.
Delivery information: CarboxyLink Columns, 2 mL, 5 columns, EDC Crosslinker, 5 x 60 mg, MES Buffered Saline Pack (makes 500 mL), 1 pack, Wash Solution, 120 mL, Column Accessories
Caution: For Research Use Only. Not for use in diagnostic procedures. Store EDC desiccated at -20˚C. Store other components at 4˚C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty