Would you like to log in again?
Pierce Eggcellent™ Chicken IgY Purification Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
Eggcellent™
44922 44918 44918 44922
PI44922EA 1300.52 USD InStock
CAPI44918 PI44922 CAPI44922 PI44918
Pierce Eggcellent™ Chicken IgY Purification Kits, Thermo Scientific
Protein Purification Protein Purification Kits
Chicken IgY can be collected from egg yolks; however, classical affinity chromatography methods such as protein A and protein G cannot be used to purify the IgY protein because of the high lipid content in the egg yolk.
  • Purifies twice the amount of IgY and obtain 80 to 120 mg IgY per egg yolk at 85% to 95% purity
  • Simple precipitation method purifies chicken IgY without affinity columns
  • Eggs can be stored in buffer and purified at a later date
These kits contain a delipidation reagent that separates the proteins from the lipids and allows the proteins to be stored in the freezer for up to one year. The protocol allows for eggs to be used immediately out of the refrigerator or stored in buffer for later purification. The precipitation works without affinity columns, yet still delivers purity levels of 85–95%.
Delivery information: Kit includes Delipidation reagent (500 mL or 5x500 mL), IgY Precipitation reagent (500 mL or 5x500 mL), Egg separator (1 or 5 separators).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty