Would you like to log in again?
Pierce™ DyLight™ PEGylated Dyes, Antibody Labelling Dyes, Thermo Scientific
Supplier: Thermo Fisher Scientific
DyLight™
62292 46601 46627 62274 62277 62274 62269 46604 62277 62294 62294 46627 46604 62269 62292 46601
PI62292EA 385 USD InStock
PI62292 PI46604 PI46601 CAPI62269 PI62277 PI62294 CAPI46627 PI62274 CAPI46604 CAPI62294 CAPI46601 CAPI62292 PI62269 CAPI62277 CAPI62274 PI46627
Pierce™ DyLight™ PEGylated Dyes, Antibody Labelling Dyes, Thermo Scientific
Protein Linking and Labeling Systems
DyLight™ PEGylated dyes display absorption spectra ranging from 550 nm to 783 nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor® or Cy® dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
The PEGylated DyLight™ dyes are derivatives of corresponding high-performance non-PEGylated DyLight™ dyes, which are used to fluorescently label antibodies and proteins
These DyLight™ dyes contain two to four non-toxic polyethylene glycol (PEG) chains. The PEG chains enhance fluorescence and reduce non specific binding of conjugates, improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability, and improve retention, notably in tumours. The near-infrared (NIR) to far-red fluorescence properties of some of the PEGylated dyes such as DyLight 680-4xPEG and DyLight 755 make them useful in biological, chemical and pharmaceutical applications, including in vivo imaging.
The amine-reactive dyes contain N-hydroxysuccinimide (NHS) esters, the simplest and most commonly used reactive group for labeling proteins. NHS esters react with primary amines on the protein, forming a stable, covalent amide bond and releasing the NHS group.
The sulphhydryl-reactive fluors contain maleimide groups that react predominantly with free –SH groups at pH 6.5 - 7.5, forming a stable thioether bond. At pH 7, the maleimide group is
Conjugates made with PEGylated DyLight™ dyes are effective molecular probes that can be used in cellular imaging and other fluorescence detection methods.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty