Would you like to log in again?
Pierce™ Marfey's Reagent, Thermo Scientific
Supplier: Thermo Scientific
Pierce
48895
PI48895EA 174.46 USD InStock
PI48895
Pierce™ Marfey's Reagent, Thermo Scientific
Assays Protein Assays
Thermo Scientific Pierce Marfey's Reagent (FDAA) reacts with primary amines to enable the quick and easy separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.
  • For precolumn derivatization of primary amines for detection by UV absorbance (λmax = 340nm)
  • Provides nanomole sensitivity in typical HPLC applications with amino acids
  • Reaction products enable chiral separation of D- and L-amino acids
  • Following reverse phase HPLC, derivatives have an extinction coefficient of 3×104 M-1 cm-1
  • Store product at room temperature
FDAA is 1-fluoro-2-4-dinitrophenyl-5-L-alanine amide, also called Marfey's reagent
FDAA derivatives can be separated with simple linear gradient of triethylamine phosphate/acetonitrile on a Spheri-5 RP-18 HPLC cartridge column. FDAA reduces the need for a chiral column because the D-derivatives are retained on the column, eluting after the L-derivatives. The derivatization is easy to perform and completed in 90 minutes, resulting in derivatives that are stable for at least 48 hours.
FDAA derivatives of D-amino acids exhibit strong intramolecular bonding, which reduces their polarity relative to the corresponding L-amino acid derivatives. Consequently, the D-derivatives are selectively retained on reverse phase columns and elute much later than corresponding L-derivatives.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty