Would you like to log in again?
TRI-SIL TBT 10 X 1 ML AMPULES
Supplier: Thermo Scientific
Tri-Sil TBT^ Group: Tri-Silr TBT. TMSI, BSA, TMCS (3:3:2) catalyzed silylation reagent formulation that converts all classes of hydroxyl groups to TMS ethers.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TRI-SIL TBT 10 X 1 ML AMPULES PI49016 TS-49016 Each (1ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty