Would you like to log in again?
Pierce™ NHS-Fluorescein Antibody Labeling Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
53029 53029
PI53029EA 537.6 USD InStock
PI53029 CAPI53029
Pierce™ NHS-Fluorescein Antibody Labeling Kit, Thermo Scientific
Protein Linking and Labeling Systems
Pierce™ NHS-Fluorescein Antibody Labeling Kit is a multi-component kit containing a high-performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.
  • Fluorescently label and purify protein in approximately one hour
  • Amine specific labeling– the isothiocyanate variety of fluorescein efficiently labels antibodies and other purified proteins at primary amines
  • Single-use vials of reagent for added convenience
Delivery information: Standard kit includes NHS Fluorescein (3×50 µg vials), borate buffer (1 mL), spin columns (6 each), collection tubes (12 each) and purification resin (5 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty