Would you like to log in again?
Pierce™ UltraLink™ Biosupport, Affinity Chromatography Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53110 53111 53110 53111
PI53111EA 1183.75 USD InStock
CAPI53111 CAPI53110 PI53111 PI53110
Pierce™ UltraLink™ Biosupport, Affinity Chromatography Resin, Thermo Scientific
Protein Purification Protein Purification Resins
UltraLink™ Biosupport is a durable, azlactone-activated, beaded-polyacrylamide resin to covalently immobilize proteins and other primary amine biomolecules for preparation of affinity purification columns.
  • With excellent reusability
  • Immobilizes proteins with very high efficiency and coupling capacity in 1 hour
  • Porous beads have a 60 µm diameter, can withstand 6,9 bar and allow for linear flow-through rates of 3000 cm/hour
UltraLink™ Biosupport is an alternative to CNBr- and NHS-activated agarose resins for immobilizing antibodies and other proteins to prepare affinity columns
The azlactone groups react rapidly and efficiently with primary amines (eg. side chain of lysine residues) to covalently immobilize protein or other amine-containing ligands. The bead structure and efficient coupling chemistry of UltraLink™ Biosupport results in high protein binding capacity, high linear flow rates, low non specific binding and overall superior performance in affinity chromatography. UltraLink™ resins are ideal for medium pressure applications such as FPLC.
The beaded resin is a hydrophilic copolymer of polyacrylamide and azlactone having a rigid polymeric structure with high surface area and pore volume.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty