Would you like to log in again?
Pierce™ Streptavidin UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53114 53114 53113 53113
PI53114EA 785.72 USD InStock
CAPI53113 CAPI53114 PI53114 PI53113
Pierce™ Streptavidin UltraLink™ Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Streptavidin UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilized onto a beaded acrylamide support
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Streptavidin UltraLink™ resin binding capacity: >2 mg of biotinylated BSA/mL of resin
UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty