Would you like to log in again?
Pierce™ UltraLink™ Immobilized Protein G, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53126 53125 53125 53127 53128 53126 53128 53127
PI53128EA 602.58 USD InStock
CAPI53125 PI53128 PI53127 PI53126 PI53125 CAPI53126 CAPI53127 CAPI53128
Pierce™ UltraLink™ Immobilized Protein G, Thermo Scientific
Protein Purification Protein Purification Resins
Protein G UltraLink™ Resin is an extremely durable beaded polyacrylamide support to which recombinant Protein G has been immobilized for use in IgG purification methods
  • Immobilized Protein G – specifically binds IgG from several mammalian species, especially all isotypes of IgG from human, mouse and rat
  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
It is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods.
Delivery information: Available as resin or as Protein G UltraLink Columns (2 x 2 mL resin pre-packed in columns with twist-off bottom tabs and an accessory pack containing two white tips).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty