Would you like to log in again?
Pierce™ Protein A/G and A/G Plus UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53135 53133 53133 53135 53132 53132
PI53135EA 646.89 USD InStock
CAPI53133 CAPI53135 CAPI53132 PI53135 PI53133 PI53132
Pierce™ Protein A/G and A/G Plus UltraLink™ Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Protein A/G UltraLink™ and Protein A/G Plus UltraLink™ resins are extremely durable beaded polyacrylamide supports to which recombinant Protein A/G has been immobilized for use in IgG purification methods
  • Specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
  • Acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods
Immobilized proteins are covalently coupled to an agarose support to provide an efficient method of purifying antibodies.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty