Would you like to log in again?
Pierce™ UltraLink™ Monomeric Avidin Resin, Affinity Chromatography Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53146 53146
PI53146EA 850.34 USD InStock
CAPI53146 PI53146
Pierce™ UltraLink™ Monomeric Avidin Resin, Affinity Chromatography Resin, Thermo Scientific
Protein Purification Protein Purification Resins
UltraLink™ Monomeric Avidin resin is ideal for affinity purification of biotinylated proteins, peptides and other molecules
  • Non-denaturing purifies biotinylated products using mild elution conditions (2 mM free biotin)
  • Reusable at least 10 times, with marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)
  • Retains biotin-binding specificity and low non specific binding of native avidin
  • Binding capacity is >1.2 mg biotinylated BSA/mL resin
Immobilization of avidin monomers results in a purification resin with a much lower biotin-binding affinity than native tetrameric avidin, enabling recovery of biotinylated molecules using mild elution conditions.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty