Would you like to log in again?
Pierce™ UltraLink™ Iodoacetyl Resin, Affinity Purification Resins, Thermo Scientific
Supplier: Thermo Fisher Scientific
UltraLink™
53155 53155
PI53155EA 400.62 USD InStock
CAPI53155 PI53155
Pierce™ UltraLink™ Iodoacetyl Resin, Affinity Purification Resins, Thermo Scientific
Protein Purification Protein Purification Resins
UltraLink™ Iodoacetyl Resin is a durable, porous resin that is activated to enable efficient and direct covalent immoblization of peptides and other ligands through their sulfhydryl groups (-SH) for use in affinity purification procedures.
  • High coupling efficiency and capacity
  • Specific and leak-proof coupling chemistry
  • Durable, high-performance resin
  • Prepared affinity resin can be used with typical binding and elution procedures for many cycles of affnity purification
The bead structure and efficiently coupling chemistry of the Iodoacetyl-activated UltraLink™ support results in high protein binding capacity, high linear flow rates, low non specific binding and overall superior performance in affinity chromatography. UltraLink™ resins are ideal for medium pressure applications, such as FPLC.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty