Would you like to log in again?
Pierce™ DyLight™ Antibody Labelling Dyes, Thermo Scientific
Supplier: Thermo Fisher Scientific
DyLight™
46414 46621 62265 46419 46622 46613 46602 46608 46613 62295 46402 62279 46412 46400 62262 46421 62295 46618 46417 62263 62266 46401 62265 46413 62263 46402 62266 46427 46600 46608 46419 46621 46413 46403 62278 46602 46403 62290 46418 46422 62290 46600 62262 46422 46400 46414 46401 46412 46418 46426 62298 46421 46618 46417
PI46621EA 520.46 USD InStock
CAPI46608 CAPI46613 CAPI46414 CAPI62262 CAPI46413 CAPI46412 CAPI62265 CAPI62266 CAPI62263 PI46621 PI62290 PI46622 PI46421 PI46422 PI46427 PI46426 PI62298 CAPI46618 CAPI46419 PI62295 CAPI46418 CAPI46417 CAPI62295 CAPI46621 CAPI46422 CAPI46421 PI46618 PI46418 PI46419 CAPI62290 PI46412 PI46413 PI46417 PI46414 PI46613 PI62265 PI62266 PI62262 PI62263 PI46608 PI46401 PI46402 PI46600 PI46400 PI46403 PI46602 PI62278 PI62279 CAPI46602 CAPI46403 CAPI46402 CAPI46600 CAPI46401 CAPI46400
Pierce™ DyLight™ Antibody Labelling Dyes, Thermo Scientific
Protein Linking and Labeling Systems
DyLight™ Dyes have absorption spectra ranging from 350 nm to 770 nm and match the output wavelengths of common fluorescence instrumentation
  • Available in both amine- and sulfhydryl-reactive chemistries for fast and efficient labeling of IgG or other proteins
  • High water solubility
  • Excellent photostability
  • Compatible with common fluorescence instrumentation
Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
The DyLight™ dyes exhibit higher fluorescence intensity and photostability than Alexa Fluor®, CyDye® and LI-COR dyes in many applications and remain highly fluorescent over a broad pH range (pH 4 to 9). Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
Delivery information: Supplied as 1 mg or in convenience packs containing sufficient reagent for 5 single labelling reactions. Each single-use vial is sufficient to label 0.25 to 1 mg of protein (>45 kDa).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty