Would you like to log in again?
Pierce™ DyLight™ Fluor-Free Acids, Antibody Labelling Dyes, Thermo Scientific
Supplier: Thermo Fisher Scientific
DyLight™
62280
PI62280EA 440 USD InStock
PI62280
Pierce™ DyLight™ Fluor-Free Acids, Antibody Labelling Dyes, Thermo Scientific
Protein Linking and Labeling Systems
DyLight™ fluor-free acids are non-activated forms of selected DyLight™ fluorescent dyes for use as controls and calibration factors in fluorescence imaging applications
These non-activated fluorescent dyes contain the native carboxyl group of the standard dye molecule.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty