Would you like to log in again?
Pierce™ Imject™ Bovine Serum Albumin (BSA)
Supplier: Pierce Chemical
Imject®
77110 77110
PI77110EA 398.46 USD InStock
PI77110 CAPI77110
Pierce™ Imject™ Bovine Serum Albumin (BSA)
Proteins and Peptides
Thermo Scientific Imject BSA in PBS is purified bovine serum albumin carrier protein that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
  • High-yield conjugation—each molecule of 59 lysine residues, 30 to 35 of which have primary amines that are capable of reacting with a conjugation reagent
  • Validated quality—purified BSA (Fraction V) is available lyophilized and stabilized in PBS or MES buffer, ready for use in glutaraldehyde, NHS-ester or EDC crosslinking methods
  • Compatible solubility—numerous carboxyl groups account for the net negative charge (pI 5.1) of BSA and its high solubility, allowing its use in a broad range of conjugation conditions
  • Immunogenic—at 67 kDa, BSA is sufficiently large to function as the primary immunogen or it can be used as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
BSA is a popular immunogenic carrier protein used for preparation of peptide antigens for immunization and antibody production. BSA preparations in phosphate buffer are ready for hapten-carrier conjugation via amine-reactive NHS-ester or glutaraldehyde crosslinking.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty