Would you like to log in again?
Pierce™ Imject™ Bovine Serum Albumin (BSA), Maleimide-activated
Supplier: Pierce Chemical
Imject®
77115 77116 77115 77116
PI77116EA 213.85 USD InStock
CAPI77115 CAPI77116 PI77116 PI77115
Pierce™ Imject™ Bovine Serum Albumin (BSA), Maleimide-activated
Proteins and Peptides
Thermo Scientific Imject Maleimide-Activated BSA enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunization and antibody production.
  • Optimized activation
  • Sulfhydryl-reactive
  • Simple, rapid procedure
  • High-quality carrier protein
  • Immunogenic and soluble
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty