Would you like to log in again?
Pierce Imject™ Chicken Ovalbumin (from Egg), Thermo Scientific
Supplier: Thermo Scientific
Pierce
77120
PI77120EA 398.46 USD InStock
PI77120
Pierce Imject™ Chicken Ovalbumin (from Egg), Thermo Scientific
Proteins and Peptides
Imject™ Ovalbumin is a purified ovalbumin (OVA) protein conveniently formulated for preparation of hapten-carrier conjugates.
  • Purified from chicken egg whites and lyophilized in phosphate buffered saline (PBS)
  • Single polypeptide protein with molecular mass of 45,000 Da
  • Contains 20 lysine, 14 aspartic acid and 33 glutamic acid residues
  • Not as immunogenic as keyhole limpet haemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent
Ovalbumin is useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty