Would you like to log in again?
Pierce Imject™ Maleimide-Activated Ovalbumin, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
77125 77126 77125 77126
PI77126EA 244.62 USD InStock
CAPI77126 CAPI77125 PI77126 PI77125
Pierce Imject™ Maleimide-Activated Ovalbumin, Thermo Scientific
Protein Extraction Protein Extraction Reagents
Imject™ Maleimide-activated ovalbumin is sulfhydryl-reactive ovalbumin (OVA) for hapten-carrier conjugation with cysteine-containing peptides and other thiolated antigens.
  • Ideal for conjugating cysteine-containing peptide antigens to make immunogen or screening target
  • Conjugation reaction complete in 2 hours; results in stable, covalent thioether bonds
  • Single polypeptide protein with molecular mass of 45000 Daltons
  • Contains 14 aspartic acid and 33 glutamic acid residues whose carboxyl groups account for the net negative charge (pI 4.63) of OVA
Imject™ Ovalbumin is not as immunogenic as other proteins, such as keyhole limpet hemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent. It is useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty