Would you like to log in again?
Imject™ Freund's Complete Adjuvant (FCA), Thermo Scientific
Supplier: Thermo Scientific
Imject®
77140 77140
PI77140EA 213.85 USD InStock
PI77140 CAPI77140
Imject™ Freund's Complete Adjuvant (FCA), Thermo Scientific
Protein Expression Protein Expression Reagents
Thermo Scientific Imject Freund's Complete Adjuvant is a popular nonspecific stimulator of the immune response for use in antibody production immunization protocols.
  • Also called Complete Freund's Adjuvant (CFA)
  • Water-in-oil emulsion containing killed Mycobacterium
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Disadvantages: difficult to mix with immunogen and can cause tissue necrosis at injection site
This product comes in 10 mL ampules that are convenient and ready to use
Adjuvants are nonspecific stimulators of the immune response. When mixed with an antigen or immunogen, adjuvants help to deposit or sequester the injected material thereby helping to increase antibody response. Adjuvants enhance the immune response to compounds that are already immunogenic; they do not confer immunogenicity to non-immunogenic haptens. To make prospective antigens more immunogenic, it is necessary to conjugate them to a carrier protein or some other complex, immunogenic molecule.
The water-and-oil emulsion greatly enhances immune responses to immunogens when mixed and injected with the prepared antigen, and is typically used for initial injections. The ampules comprise conveniently sized aliquots that provide long shelf life.
Ordering information: Upon receipt store at 4?C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty