Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Imject™ Freund's Incomplete Adjuvant (FIA), Thermo Scientific

Supplier: Thermo Scientific
Imject®
77145 77145
PI77145EA 198.46 USD InStock
CAPI77145 PI77145
Imject™ Freund's Incomplete Adjuvant (FIA), Thermo Scientific
Protein Expression Protein Expression Reagents
Thermo Scientific Imject Freund's Incomplete Adjuvant is a popular nonspecific stimulator of the immune response for use in antibody production immunization protocols.

  • Water-in-oil emulsion without Mycobacterium
  • Enhances immune response in booster injections
  • Typically used for subsequent boosts after initial injection with FCA
  • Disadvantage: difficult to mix with immunogen

This product comes in 10 mL ampules that are convenient and ready-to-use

Adjuvants are nonspecific stimulators of the immune response. When mixed with an antigen or immunogen, adjuvants help to deposit or sequester the injected material thereby helping to increase antibody response. Adjuvants enhance the immune response to compounds that are already immunogenic; they do not confer immunogenicity to non-immunogenic haptens. To make prospective antigens more immunogenic, it is necessary to conjugate them to a carrier protein or some other complex, immunogenic molecule.

The water-and-oil emulsion greatly enhances immune responses to immunogens when mixed and injected with the prepared antigen, and is typically used for booster injections. The ampules comprise conveniently sized aliquots that provide long shelf life.

Ordering information: Upon receipt store at 4 °C.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty