Would you like to log in again?
Pierce™ Imject™ Bovine Serum Albumin (BSA), Cationized, Pierce Chemical
Supplier: Pierce Chemical
Imject®
77150 77150
PI77150EA 676.92 USD InStock
PI77150 CAPI77150
Pierce™ Imject™ Bovine Serum Albumin (BSA), Cationized, Pierce Chemical
Proteins and Peptides
Thermo Scientific Imject Cationized Bovine Serum Albumin (cBSA) functions as an easy-to-use and highly immunogenic carrier protein for preparing antigens for use in antibody production.
  • Purified bovine serum albumin (BSA) that is cationized and supplied lyophilized in PBS
  • Single polypeptide protein with molecular mass >67,000 Daltons
  • Contains 59 lysine residues, plus numerous other primary amines added by cationization, most of which are capable of reacting with a conjugation reagent to attach antigens
  • Net positive charge (pI >11), conferring solubility in conditions not suitable for the negatively-charged native BSA (pI 5.1)
  • Significantly greater immunogenicity than native BSA because the positively charged protein is processed differently by the immune-response system
Imject cBSA is prepared by modifying native BSA with excess ethylenediamine, essentially capping all negatively-charged carboxyl groups with positively-charged primary amines.
Imject cBSA is an excellent choice for immunogen preparation when greater immunogenicity is needed relative to BSA but conditions are not amenable to using keyhole limpet hemocyanin (KLH) or Blue Carrier Protein.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty