Would you like to log in again?
Pierce™ Imject™ Blue Carrier™ Immunogenic Proteins, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
77130 77660
PI77130EA 329.45 USD InStock
PI77130 PI77660
Pierce™ Imject™ Blue Carrier™ Immunogenic Proteins, Thermo Scientific
Protein Extraction Protein Extraction Kits
Pierce™ Imject™ Blue Carrier™ protein is a highly soluble, mollusc-derived hemocyanin that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
  • High-yield conjugation with each molecule of CCH contains hundreds of primary amines for coupling haptens via EDC or NHS ester crosslinkers
  • Validated quality for the immunogenic carrier protein to ensure uniform chemical structure and solubility
  • Superior solubility and highly immunogenic
The two subunits form an extremely stable heterodidecameric structure, even in the absence of divalent cations. The arrangement of the subunits contains diverse repeated antigenic structures that elicit a strong immune reaction from B and T lymphocytes. This carrier protein can be used in any application requiring flexibility in the choice of buffer and pH.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty