Would you like to log in again?
Imject™ Pierce™ Maleimide-Activated Carrier Proteins, Thermo Scientific
Supplier: Thermo Fisher Scientific
Imject®
77606 77610 77663 77610 77605 77605 77606
PI77606EA 198.46 USD InStock
CAPI77610 PI77606 PI77605 PI77610 CAPI77605 CAPI77606 PI77663
Imject™ Pierce™ Maleimide-Activated Carrier Proteins, Thermo Scientific
Protein Extraction Protein Extraction Kits
Imject® Maleimide-activated mcKLH and PEGylated mcKLH enable conjugation of sulfhydryl-containing peptide haptens to elicit an immune response and antibody production against the hapten
  • Used to achieve the highest antigen conjugation efficiency
  • Cysteine reactive chemistry enables controlled orientation of your antigen
  • High solubility minimises loss during conjugation
The mcKLH is maleimide-activated using a heterobifunctional crosslinker that contains an N-hydroxysuccinimide (NHS) ester and a maleimide group. The PEGylated mcKLH is activated with NHS-PEG2-Maleimide, creating a more soluble maleimideactivated mcKLH. Maleimide groups react with free sulfhydryls, forming stable thioether bonds.
Ordering information: Upon receipt store product at −20 °C. Product is shipped with an ice pack.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty