Would you like to log in again?
Imject™ EDC Carrier Protein Spin Kits, Thermo Scientific
Supplier: Thermo Scientific
Imject®
77672 77670 77671 77671
PI77671EA 1015.38 USD InStock
PI77671 PI77670 CAPI77671 PI77672
Imject™ EDC Carrier Protein Spin Kits, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Imject™ EDC Carrier Protein Spin Kits enable effective one-step conjugation of a hapten to a carrier protein using EDC as the crosslinker. The resulting conjugate is used for eliciting an immune response and antibody production against the hapten.
EDC reacts with exposed carboxyl and amino groups on peptides and proteins to form stable bonds
Blue Carrier™ Protein is a haemocyanin from the Concholepas concholepas mollusc with immunogenic properties similar to KLH but is more stable and efficient as a carrier protein for the production of antibodies to haptens and peptides. Mariculture keyhole limpet haemocyanin (mcKLH) produces an excellent immune response and is currently the industry standard for antibody production against a hapten or peptide. mcKLH is harvested from the haemolymph of giant keyhole limpets (Megathura crenulata) maintained in environmentally optimized marine tanks and are not sacrificed during the haemolymph collection. Bovine serum albumin (BSA) constitutes approximately half of the protein in plasma and is often used as a non relevant carrier in an ELISA to assess anti-peptide antibody titres.
These kits contain carrier proteins formulated in buffers compatible with EDC reactions and Zeba™ Desalt Spin Columns, which offer exceptional desalting and protein recovery by centrifugation eliminating the waiting and monitoring associated with gravity-flow columns.
Ordering information: All kits contain sufficient materials for five conjugation reactions. All kits include Imject™ EDC Conjugation Buffer, 30 mL; EDC (1-ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride), 5 x 10 mg; Imject™ Purification Buffer Salts, 5 x 0.84 g; and Zeba™ Spin Desalting Columns, 5 x 2 mL.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty