Would you like to log in again?
Pierce™ TCEP Solution, Neutral pH, Bond-Breaker™, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
77720 77720
PI77720EA 249.29 USD InStock
CAPI77720 PI77720
Pierce™ TCEP Solution, Neutral pH, Bond-Breaker™, Thermo Scientific
Protein Purification Protein Purification Reagents
Bond-Breaker™ TCEP Solution (neutral pH) is a stable, 0.5 M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulphide bonds.
  • Odourless
  • Selective and complete reduction of even the most stable water-soluble alkyl disulfides
  • Reduces protein disulfides at room temperature and pH 5 in less than five minutes
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups found in proteins
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty