Would you like to log in again?
Leupeptin Protease Inhibitor
Supplier: Pierce Biotechnology
78435 78435
PI78435EA 505.11 USD InStock
CAPI78435 PI78435
Leupeptin Protease Inhibitor
Proteins and Peptides
Leupeptin Protease Inhibitor reversably inhibits serine and cysteine proteases.
  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Reliable supplement
Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty