Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ Glutathione Magnetic Agarose, Thermo Scientific

Supplier: Thermo Fisher Scientific
78602 78601
PI78601EA 260.92 USD InStock
PI78601 PI78602
Pierce™ Glutathione Magnetic Agarose, Thermo Scientific
Protein Purification Protein Purification Magnetic Beads
Thermo Scientific™ Pierce™ Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST) from a bacterial, yeast, or mammalian crude cell lysate.

  • High-performance beads: Non-aggregating, magnetite (Fe3O4), superparamagnetic beads provide exceptional uniformity for both manual and automated HTS applications
  • Stable affinity ligand: Glutathione is covalently immobilized to particles, enabling leach-resistance and clean purification products
  • High capacity: Binding capacity is sufficient for both routine and demanding purification procedures

Thermo Scientific™ Pierce™ Glutathione Magnetic Agarose Beads provide a fast, convenient method for purification of glutathione-S-transferase (GST) from a bacterial, yeast, or mammalian crude cell lysate

The high-performance, magnetite, superparamagnetic particles are validated and optimized for use with high-throughput magnetic platforms, such as the KingFisher™ 96 and KingFisher™ Flex magnetic particle processors, but the beads also enable premium performance for simple benchtop purification applications using an appropriate magnetic stand.

Glutathione S-transferase (GST) is a protein that is commonly added to the end of recombinant proteins to aid in their purification or detection. To make an affinity support, glutathione is immobilized through its central carbon, preserving the essential structure necessary for efficient GST binding. Addition of reduced glutathione in a physiologic buffer is sufficient to competitively displace and recover the GST fusion protein from the immobilized glutathione. Store at 4° C. For Research use only. Not for use in diagnostic procedures.

Pierce™ Glutathione Magnetic Agarose Beads are well suited for purifying GST fusion proteins from a soluble protein extract and are ideal for the purification of proteins expressed at low levels from diluted supernatants. The beads can be used in manual applications with a magnetic stand or automated applications with an instrument such as the Thermo Scientific™ KingFisher™ Flex Magnetic Particle Processor. Automated instruments are especially useful for higher throughput purification and screening of purification conditions.

Ordering information: Accessories not included.

Packaging: 1 mL settled resin. Supplied as 4 mL of 25% v/v suspension in 20% ethanol.

Caution: Store at 2–8°C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty