Would you like to log in again?
Pierce™ APTS (3-aminopropyltriethoxysilane), Thermo Scientific
Supplier: Thermo Scientific
80370 80370
PI80370EA 179.63 USD InStock
CAPI80370 PI80370
Pierce™ APTS (3-aminopropyltriethoxysilane), Thermo Scientific
Assays Enzyme Assays
Thermo Scientific Pierce APTS (3-aminopropyltriethoxysilane) is a silylation reagent for coating glass and silica surfaces to add primary amines, which can be used to crosslink and immobilize proteins and other molecules.
  • Crosslinking agents such as BS3, EDC and Sulfo-SMCC can be used to immobilize proteins, peptides, modified DNA, etc., to the treated surface
  • Coats certain plastic surfaces noncovalently to create a reactive film
  • Chemically adheres tissue sections to glass slides for immunohistochemistry
3-Aminopropyltriethoxysilane (APTS) is a surface treatment used to chemically couple various ligands to glass or silica surfaces such as glass slides. 3-aminopropyltriethoxysilane is first coupled to glass or silica through the silane. Compounds of interest are then coupled to the newly added amino groups directly, or additional chemistries are applied using the amino function prior to coupling.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty