Would you like to log in again?
DyLight™ Antibody Labeling, Long Stokes Shift Dyes, Thermo Scientific
Supplier: Thermo Scientific
82491 82493 82495 82493 82491 82495
PI82491EA 452.4 USD InStock
PI82491 CAPI82495 PI82493 CAPI82493 CAPI82491 PI82495
DyLight™ Antibody Labeling, Long Stokes Shift Dyes, Thermo Scientific
Protein Linking and Labeling Systems
The DyLight™ Long Stokes Shift Dyes are amine-reactive fluorescent dyes that produce long Stokes (LS) shifts of 80 - 145 nm in excitation/emission wavelengths.
The DyLight™ LS dyes provide multicolour detection using a single excitation source in applications including flow cytometry, conjugation, DNA-sequencing, microscopy and fluorescence in situ hybridization (FISH). These dyes contain N-hydroxysuccinimide (NHS) esters, the most commonly used reactive group for labeling proteins. NHS esters react with primary amines on proteins and other molecules, forming a stable, covalent amide bond and releasing the NHS group.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty