Would you like to log in again?
Pierce™ DyLight™ Antibody Labeling Kits, Thermo Scientific
Supplier: Thermo Fisher Scientific
DyLight™
53057 53063 53046 53056 53021 53024 53062 84531 84538 53056 84536 53047 84536 84531 53045 53063 53025 53057 62275 53024 53021 53020 53025 53044 84535 53047 53020 53062 53046 84530 84530 53044 53045 84535 84539
PI84530EA 727.08 USD InStock
CAPI53045 CAPI53024 CAPI53046 CAPI53025 CAPI53047 CAPI53063 CAPI53020 CAPI53021 CAPI53044 CAPI53062 PI84530 PI84531 CAPI84530 PI53057 PI53056 CAPI53056 CAPI53057 CAPI84536 CAPI84535 CAPI84531 PI62275 PI53020 PI53063 PI84536 PI53062 PI84535 PI84538 PI84539 PI53047 PI53025 PI53046 PI53024 PI53045 PI53044 PI53021
Pierce™ DyLight™ Antibody Labeling Kits, Thermo Scientific
Protein Linking and Labeling Systems
The DyLight™ Antibody Labeling Kits and were specifically developed for fast, efficient labeling of antibodies
  • Fast – fluorescently label and purify protein in approximately one hour
  • Amine-reactive dyes – label virtually any protein
  • Pre-measured fluorescent dye – eliminate the time, waste and hassle associated with weighing dye
  • Efficient non-reacted dye removal
  • Minimal sample dilution
  • Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column
  • Easy protocol
Two convenient kit formats are available to accommodate varied labeling requirements. The DyLight™ Antibody Labeling Kits contain all necessary components to perform three separate labeling reactions using 1mg of IgG or similar quantities of other proteins. The DyLight™ Microscale Antibody Labeling Kits contain all the necessary components to perform five separate labeling reactions using 100 μg of IgG. The labeling kits use high-performance spin desalting columns to provide exceptional dye removal and antibody recovery.
Ordering information: Standard kit includes Reaction Buffer (1 mL), spin columns (6 each), microcentrifuge collection tubes (12 each) and purification resin (5 mL). Each Microscale kit includes Reaction Buffer (1 mL), spin columns (5 each), microcentrifuge collection tubes (10 each) and purification resin (5 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty