Would you like to log in again?
Pierce™ Western Blotting Filter Paper, Thermo Scientific
Supplier: Thermo Scientific
Pierce
88600 84784 84783 84784 84783 88600
PI84783EA 95.06 USD InStock
CAPI88600 CAPI84784 CAPI84783 PI84783 PI88600 PI84784
Pierce™ Western Blotting Filter Paper, Thermo Scientific
Blotting Paper
Use to efficiently transfer protein from an electrophoresis gel to a membrane in northern, Southern, and Western blotting applications.
  • High-quality
  • Easy to use
  • Two thicknesses
  • Tested and recommended for use with various protein methods
Filter paper for Western blotting applications is available in various formats including precut sizes that match standard gel sizes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty