Would you like to log in again?
SuperSignal™ Enhanced Molecular Weight Protein Ladder
Supplier: Pierce Chemical
SuperSignal®
84786 84786
PI84786EA 386.18 USD InStock
PI84786 CAPI84786
SuperSignal™ Enhanced Molecular Weight Protein Ladder
Proteins and Peptides
Thermo Scientific SuperSignal Enhanced Molecular Weight Protein Ladder (20 to 150K) are a ready-to-use mix of recombinant proteins with IgG-binding sites for chemiluminescent, fluorescent, chromogenic or other detection systems.
Features of SuperSignal Enhanced Molecular Weight Protein Ladder:
Compatible—enhanced version for compatibility with mouse monoclonal primary antibodies
Ready-to-use—liquid format is ready to load and requires no boiling
Direct visualization—see marker bands with gel stains and blotting detection reagents
Consistent—even band development regardless of detection method
Stable—store long term at -20˚C, three months at 4˚C or one month at ambient
SuperSignal Enhanced Molecular Weight Protein Ladder contain ready-to-use stabilized mixtures of eight recombinant proteins ranging in size from 20 to 150kDa. These recombinant proteins bind antibodies used in the Western blot through an IgG binding site. The protein markers can then be visualized either using appropriate substrates for enzyme-labeled antibodies or via fluorescent dye-labeled antibodies. The MW marker mixes also contain a special pink tracking dye to monitor the electrophoresis run and membrane transfer without the need for additional colored markers. The standards are compatible with nitrocellulose and PVDF membranes, multiple blocking buffers and many other detection methods.
SuperSignal Molecular Weight Protein Ladders provide reliable and proportional band intensities in stained gels and immunoblots developed with chemiluminescent, fluorescent, chromogenic or other detection systems. Two versions of the MW marker are offered. The regular formulation is appropriate for use with most rabbit and other non-mouse polyclonal antibodies. The enhanced ladder is formulated specifically for applications requiring mouse monoclonal antibodies and experiments where very dilute antibody concentrations are used. For example, the enhanced standard may be necessary when using a 1/50,000 (25 ng/mL) to 1/100,000 (10 ng/mL) dilution of secondary antibody with SuperSignal West Pico Chemiluminescent substrate.
Ordering information: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty