Would you like to log in again?
Pierce™ C18 Spin Tips, Thermo Scientific
Supplier: Thermo Fisher Scientific
84850 84850
PI84850PK 398.77 USD InStock
CAPI84850 PI84850
Pierce™ C18 Spin Tips, Thermo Scientific
Protein Extraction Protein Extraction Reagents
The C18 Spin Tips enable fast and efficient capture, concentration, desalting, and elution of up to 10 µg peptides per 20 µL sample for mass spectrometry (MS) analysis.
  • Rapid—C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
  • High capacity—up to 10 µg peptide per 20 µL solution
  • Convenient—spin tips come with tip adaptors for easy centrifugation
  • Cleaner sample—device design filters out particulates that can cause autosampler and column clogging
Pierce C18 Spin Tips are 20 µL-capacity pipette tips with accompanying adaptors for microcentrifuge sample processing
Pierce C18 Spin Tips offer excellent flow properties with a high-efficiency C18 sorbent for fast wetting, loading, washing and eluting. Sample is simply loaded in prepared tip and washed and eluted using multiple centrifugation steps. The procedure is simple and requires less than 5 minutes to process protein digests, strong cation exchange fractions and other protein and peptide samples for mass spectrometric (MS) analyses. The unique tip design also removes any particulates in the sample, eliminating clogging of the auto sampler or column upstream of the mass spectrometer.
The tips contain a C18 reversed-phase sorbent that minimizes flow resistance and provides excellent binding and recovery characteristics at a wide range of peptide concentrations upstream of matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. Sample clean-up with C18 resin significantly improves protein analysis results by removing urea, salts, and other contaminants before mass spectrometry (MS). Each Pierce C18 Spin Tip has a 20 µL volume capacity with a 10 µg peptide-binding capacity.
Ordering information: Store at room temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty