Would you like to log in again?
Pierce™ Albumin Depletion Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
85160 85160
PI85160PK 464 USD InStock
CAPI85160 PI85160
Pierce™ Albumin Depletion Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
Pierce™ Albumin Depletion Kit improves serum component analysis by rapidly removing abundant albumin protein from serum samples
  • Improves performance and eases processing of multiple samples
  • Can be adapted to larger or smaller columns or 96-well filter plates
  • Removing excess albumin facilitates analysis of low-abundance serum proteins
  • Binding capacity of 2 mg of human serum albumin (HSA) for 200 µl resin
Each aliquot of 200 µl settled resin can process 5 to 50 µl of serum sample in less than 10 minutes by low-speed centrifugation using the Pierce™ spin columns provided in the kit. The kit has been optimised for human serum albumin (HSA), but also effectively binds swine, sheep and rabbit serum albumin. With modification of the binding buffer, Pierce™ albumin depletion resin can bind bovine, calf, goat and rat albumin; however, this product is not suitable for mouse albumin
The albumin depletion resin provided in the kit is a high capacity, immobilized Cibacron Blue dye agarose resin, which binds a variety of species-specific albumins.
Delivery information: Kit includes 10 ml resin slurry, 10 ml binding-wash buffer, and 24 microcentrifuge spin columns.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty