Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water with 0.1% (v/v) Trifluoroacetic acid for LC-MS, Pierce™

Supplier: Thermo Fisher Scientific

85173 T85172-K2 T85172-K2 85172
PI85172EA 112.51 USD InStock
PI85172 PI85173 CAPI85173 CAPI85172
Water with 0.1% (v/v) Trifluoroacetic acid for LC-MS, Pierce™
Water with 0.1% (v/v) Trifluoroacetic acid
Pierce™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade is a preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

  • Tested more than 20 ways to ensure maximal LC-MS sensitivity
  • Processed, filtered and sealed to extend LC-MS column life
  • Packaged to eliminate variability and reduce handling

0.1% Trifluoroacetic Acid (TFA) in water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography- and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

For complex separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve separations that would otherwise overlap. TFA is the most frequently used modifier for separations in reverse-phase HPLC. The usual specified TFA concentration is 0.1%. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make TFA concentrations the same.

Caution: For Research Use Only. Not for use in diagnostic procedures.


Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.5351 g/cm³ (20 °C)
Storage Temperature: Ambient
ADR: 8,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/92/728392.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty