Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Proline, lyophilized powder for SILAC

Supplier: 0000013509
Synonyms: (S)-(-)-ProlineH-Pro-OHL-ProlineH-Pro-OH

88211 88211
PI88211EA 64.52 USD InStock
CAPI88211 PI88211
L(-)-Proline, lyophilized powder for SILAC
L(-)-Proline
The Pierce™ Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits.

  • 100% label incorporation into proteins of living cells
  • High-quality supplements—heavy amino acids with>99% isotope purity

This is a lyophilized preparation of L-Proline, sufficient for 1 SILAC experiment.

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of "heavy" 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
Merck Index: 12,07963
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty