Would you like to log in again?
Pierce™ HisPur™ Ni-NTA Resin, Thermo Scientific
Supplier: Thermo Fisher Scientific
HisPur™
88223 88223 88222 88222 88221 88221
PI88223EA 4343.69 USD InStock
CAPI88222 CAPI88221 CAPI88223 PI88223 PI88222 PI88221
Pierce™ HisPur™ Ni-NTA Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ HisPur™ Ni-NTA Resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • Binding up to 60 mg of 6xHis-tagged protein per milliliter of resin
  • Purify proteins using native or denaturing conditions
  • Reuse the same batch of resin at least five times
  • Pre-formulated buffers available for kit formats
The specially prepared support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni²⁺). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty