Would you like to log in again?
Pierce™ HisPur™ Ni-NTA Spin Purification Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
88228 88229 88229 88228 88227 88227
PI88229PK 499.91 USD InStock
PI88229 CAPI88228 CAPI88227 CAPI88229 PI88228 PI88227
Pierce™ HisPur™ Ni-NTA Spin Purification Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ HisPur™ Ni-NTA Spin Purification Kit contains 0.2-ml HisPur™ Ni-NTA spin columns, as well as buffers required to purify polyhistidine-tagged proteins
  • High capacity
  • Compatible
  • Cost-effective
  • Easy to use
  • Flexible
HisPur™ Ni-NTA Resin effectively purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates, such as those that are attained with Thermo Scientific B-PER Bacterial Protein Extraction Reagents. The resin performs well in batch-binding and spin-column procedures at a variety of scales. Performance equals or exceeds popular Ni-NTA resins from other suppliers. HisPur™ Ni-NTA Resin is a high-quality, stable and resilient affinity support. Tests confirm that no decrease in performance occurs after at least five repeated uses. These data indicated that the resin is highly resistant to structural degradation or nickel ion leaching during normal use.
HisPur™ Ni-NTA resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
Delivery information: Kit includes HisPur Ni-NTA spin columns, phosphate-buffered saline (10X), imidazole (2 M), collection tubes and column plugs.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty