Would you like to log in again?
pMCS Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
Supplier: Thermo Scientific
88231
PI88231EA 182.4 USD InStock
PI88231
pMCS Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
Assays Protein Assays
The Thermo Scientific™ pMCS Tluc16-DD Vector is a multiple cloning site (MCS) vector designed to accept a promoter sequence for study of gene regulation using the intracellular TurboLuc™16 (Tluc16) luciferase reporter.Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systemsMultiple unique cloning sites provide versatility for transfer of regulatory elements from one vector to anotherDual-destabilization (DD) technology reduces accumulation of Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assaySynthetic polyA terminator and Transcriptional Pause Site (TPS) included upstream of MCS to minimize non-specific transcriptional read-throughTluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family
The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression. The Tluc16 luciferase expression vector also contains dual-destabilization (DD) technology that reduces accumulation of the Tluc16 luciferase mRNA and protein in cells, enhancing the responsiveness of the assay. A synthetic polyA terminator and a Transcriptional Pause Site (TPS) are included upstream of the MCS to minimize non-specific transcriptional read-through. The Tluc16 luciferase activity can be measured using the
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Store in freezer (-5 to -30˚C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty