Would you like to log in again?
pEF1α Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
Supplier: Thermo Scientific
88241
PI88241EA 181.2 USD InStock
PI88241
pEF1α Tluc16-DD Vector for Luciferase Assays, Thermo Scientific
Assays Protein Assays
The Thermo Scientific™ pEF1α Tluc16 Vector encodes the smallest known luciferase, TurboLuc™16 (Tluc16) luciferase, as a reporter under a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in mammalian cells.Intracellular Tluc16 luciferase gene, optimized for high expression in mammalian systemsTruncated EF1α promoter for constitutive Tluc16 luciferase expressionTluc16 luciferase is a 16 kDa, novel, intracellular luciferase derived from the marine copedod Metridia luciferase family
The wild-type luciferase has been modified to reduce its size, increase its brightness, and enable its intracellular expression. The Tluc16 luciferase activity can be measured using the
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Store in freezer (-5 to -30˚C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty