Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pierce™ High Capacity Endotoxin Removal Resin, Thermo Scientific

Supplier: Thermo Fisher Scientific
Pierce
88271 88270 88271 88270 88272 88272
PI88271EA 3729.85 USD InStock
PI88271 PI88270 PI88272 CAPI88271 CAPI88270 CAPI88272
Pierce™ High Capacity Endotoxin Removal Resin, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ High Capacity Endotoxin Removal Resins are ideal for the removal of endotoxins from protein or antibody samples prior to cell transfection or animal injection.

  • Bind up to 2 000 000 EU/ml to eliminate <99% of endotoxins
  • Recover greater than 85% of protein sample

Pierce™ High Capacity Endotoxin Removal Resin is a porous cellulose bead that has been surface-modified with covalently attached polylysine chains that have a high affinity for LPS but lacks the toxicity of other endotoxin-binding molecules. With a 2,000,000 EU/ml binding capacity, endotoxin levels can be reduced by 99% in samples containing initial endotoxin levels of 10,000 EU/ml. Typical protein samples processed with the High Capacity Endotoxin Removal Resin have a final endotoxin concentration below 5 EU/ml. This resin is offered in a slurry format for custom packing of endotoxin removal columns for gravity flow or flow rates ranging from 10 to 15 ml/hour.

Delivery information: Supplied as 50% slurry in 20% ethanol

Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty