Would you like to log in again?
Pierce™ Primary Cardiomyocyte Isolation Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
88281 88281
PI88281EA 674.46 USD InStock
PI88281 CAPI88281
Pierce™ Primary Cardiomyocyte Isolation Kit, Thermo Scientific
Protein Extraction Protein Extraction Kits
The Pierce™ Primary Cardiomyocyte Isolation Kit provides isolation and culturing reagents for the optimal yield and viability of primary cardiomyocytes from neonatal heart from mouse and rat.
  • Procedure and reagents optimized for viability, yield, purity, and ease-of-use
  • Isolation protocol requires less than 2 hours compared to up to 21 hours by other methods
  • Provides a 1.5- to 4-fold increase in yield compared to do-it-yourself methods and other commercial kits
  • Provides higher viability than do-it-yourself methods and other commercial kits
  • Cultured cardiomyocytes express the appropriate biochemical markers with validated beating activity
The Pierce™ Primary Cardiomyocyte Isolation Kit has been designed to obtain high viability and purity of primary cardiomyocytes using validated reagents and an optimized procedure for enzymatic digestion and culturing. The kit contains two tissue-specific dissociation enzymes, media formulated for primary cell culture, and a cardiomyocyte cell culture growth supplement that promotes the isolation, enrichment and growth of functional cardiomyocytes. The validated protocol ensures optimal isolation and significantly reduces the processing time of isolation when compared to other methods. Addition of the cardiomyocyte growth supplement to culture cells further enriches for cardiomycytes, and cultures have been maintained for up to 7 days using the kit components.
Ordering information: Upon receipt store Product 88281X at 4?C and Product 88281Y at -20?C. Product 88281X is shipped on ice packs and Product 88281Y is shipped with dry ice.
Delivery information: Kit includes Cardiomyocyte Isolation Enzyme 1, Cardiomyocyte Isolation Enzyme 2, Hanks' Balanced Salt Solution (HBSS), D-MEM for Primary Cell Isolation, and Cardiomyocyte Growth Supplement.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty